Learn More
YMC America, Inc TriBioC18 300A 5um 50x.5mm1/16
SDP
Supplier:  YMC America, Inc TA30S0505J0AU
Encompass
YMC-Triart phases are based on organic/inorganic hybrid particles that combine the high mechanical stability and efficiency of a silica-based packing material with the robust chemical stability of a polymer-based packing material This versatile resin is produced using tightly controlled particle formation technology adapted from micro-reactor technology The resulting uniform particle and pore size distributions as well as the smooth surface greatly contribute to excellent peak shape and separation reproducibility In addition YMC-Triart materials are highly resilient and robust when used at extreme pH elevated temperature and high pressure
Catalog No. 50-008-77477