Learn More
TRIBIOSCIENCE INC Bovine QPCR, 100 reactions
SDP
Supplier:  TRIBIOSCIENCE INC TBS42001
Encompass
The Bovine QPCR Kit is designed for the target-DNA specific detection of bovine matter in food and animal feed ingredients. The assay uses a real-time PCR-based protocol with fluoresce-probe to detect target DNA.
Catalog No. NC2145493