Learn More
Medchemexpress LLC HY-W011777 500mg Medchemexpress, Tricaine (methanesulfonate) CAS:886-86-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW011777500MG
Encompass_Preferred
Medchemexpress, HY-W011777 500mg Tricaine (methanesulfonate) CAS:886-86-2 Tricaine methanesulfonate (MS-222) possesses anesthetic effect. Tricaine methanesulfonate (MS-222) is common used to immobilize fish for marking or transport and to suppress sensory systems during invasive procedures. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3317