Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tricalcium phosphate | 7758-87-4 | 98.0% | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYD0833

Encompass_Preferred

Tricalcium phosphate is a biological material or organic compound widely used in life science research. It is suitable for various laboratory applications, including biochemical assays. For optimal stability, the product should be stored at -20°C for up to 3 years or 4°C for up to 2 years in powder form. If in solvent, store at -80°C for 6 months or -20°C for 1 month.

  • Molecular formula: Ca3O8P2
  • Molecular weight: 310.18
  • Purity: 98.00%
  • Target: Biochemical assay reagents
  • Pathway: Others

Catalog No. 50-004-14092


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.