Learn More
Medchemexpress LLC Tricarbonylchloro(glycinato)ruthenium | 475473-26-8 | MFCD07364048 | 97.5% | 294.61 g/mol | C5H4ClNO5Ru | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10058110MG
Encompass_Preferred
CORM-3 is a ruthenium-based carbon monoxide (CO) releasing molecule used in biochemical and cell-based research to study CO signaling and inflammation modulation. It has been reported to attenuate NF-κB p65 nuclear translocation, reduce reactive oxygen species, and enhance intracellular antioxidant defenses such as glutathione and superoxide dismutase.
  • Water-soluble ruthenium carbonyl complex suitable for biochemical studies.
  • Releases carbon monoxide to modulate inflammatory signaling.
  • Reduces reactive oxygen species and NLRP3 inflammasome activation.
  • Enhances intracellular glutathione and superoxide dismutase levels.
  • Available in multiple pack sizes including small-mass vials for dosing flexibility.
  • Stable as powder at -20°C and in solvent under recommended storage conditions.
Catalog No. 50-002-12664