Learn More
Medchemexpress LLC Tricarbonyldichlororuthenium(II) dimer | 22594-69-0 | MFCD00011528 | 98.0% | 512.01 g·mol⁻¹ | C6H6Cl4O6Ru2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW03357710MM/1ML
Encompass_Preferred
Tricarbonyldichlororuthenium(II) dimer is a carbon monoxide-releasing molecule used as a laboratory CO donor in biochemical and pharmacological research. It is supplied as a DMSO solution or as a solid and is used to investigate CO-mediated signaling, cytoprotection, antioxidant, and anti-inflammatory effects in controlled experimental settings.
  • Releases controlled amounts of CO for signaling studies.
  • High purity suitable for research applications.
  • Soluble in DMSO for ready-to-use solutions.
  • Stable under recommended storage conditions.
  • Applicable in in vitro and in vivo experimental models.
Catalog No. 50-003-00087