Learn More
Indofine Chemical 5,7,3',4',5'-Pentahydr, 5 Mg
SDP
Supplier:  Indofine Chemical 021335S 5 MG
Encompass_Preferred
5,7,3',4',5'-PENTAHYDROXYFLAVONE with HPLC 5 MG, Flavone, Tricetin with HPLC, C15H10O7, MW: 302.24, 99% by HPLC, 520-31-0, MFCD00210581
Catalog No. 50-909-353