Learn More
Diagnostic Biosystems TRICHOME STAIN Trichome Stain
Supplier:  Diagnostic Biosystems KT034
Encompass_Preferred
The Trichrome Stain Kit (Modified Masson’s) is intended for use in the histological visualization of collagenous connective tissue fibers in formalin-fixed tissue sections.
Catalog No. 50-240-7547