Learn More
Cayman Chemical TrIchostatIn C 2.5mg
SDP
Supplier:  Cayman Chemical 202202.5 MG
Encompass_Preferred
A glycosylated derivative of trichostatin A that has been shown to induce the differentiation of a mouse erythroleukemia cell line and to increase histone H4 acetylation in B cells, though at higher concentrations than trichostatin A
Catalog No. 50-298-3578