Learn More
Alkali Scientific Tricine SDS Sample Buffer 2X Solution, 25 Milliliters
Supplier: Alkali Scientific CIN001CP
Tricine SDS Sample Buffer 2X Solution is designed for use in protein analysis, particularly in SDS-PAGE electrophoresis. The 2X concentration ensures optimal sample preparation by providing the necessary components for protein denaturation and stability during electrophoresis. Tricine, a component of this buffer, offers enhanced resolution for small molecular weight proteins compared to other buffers like Tris-Glycine. This 25mL bottle is ideal for use in routine protein analysis, enabling the preparation of protein samples for separation on SDS-PAGE gels. Its formulation ensures consistent results with sharp bands and reliable reproducibility in electrophoresis-based applications.