Learn More
Cayman Chemical TrIcIrIbIn Phosphate 10mg
SDP
Supplier:  Cayman Chemical 3798510 MG
Encompass_Preferred
An Akt activation inhibitor and active metabolite of triciribine; binds to the PH domain of Akt and prevents its recruitment to the plasma membrane in MDA-MB-468 cells; reduces the proliferation of A1847 and A2780 cells (IC50s = 1.6 and 18 µM, respectively); inhibits HIV-1 reverse transcriptase activity in CEM-SS cells (IC50 = 0.04 µM), as well as HSV-1 replication in BS-C-1 cells and human CMV plaque formation in HFF-1 IRR cells (IC50s = 20 and 0.8 µM, respectively)
Catalog No. 50-291-7671