Learn More
Medchemexpress LLC Triciribine (API-2) | 35943-35-2 | C13H16N6O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1545710MM/1ML
Encompass_Preferred
Triciribine is a nucleoside analog and purine analog that acts as a DNA synthesis inhibitor. It also selectively inhibits the phosphorylation and activation of all three Akt isoforms, with IC50 values of 130 nM for Akt and 0.02-0.46 μM for HIV-1/2. This product is for research use only.
  • Inhibits DNA synthesis
  • Inhibits Akt with an IC50 of 130 nM (cell assay)
  • Inhibits HIV-1 and HIV-2 with IC50 values of 0.02-0.46 μM
  • Selectively inhibits phosphorylation and activation of all three Akt isoforms
  • Inhibits Akt phosphorylation at Thr308 and Ser473 at 10 μM concentration
  • Effectively inhibits phosphorylation and catalytic activity of Akt in PC-3 cells
Catalog No. 50-003-76659