Learn More
Medchemexpress LLC Triciribine phosphate sodium | 99.8% | 422.27 g·mol^-1 | C13H16N6NaO7P | 25 MG

Supplier: Medchemexpress LLC HY10250A25MG
Triciribine phosphate sodium is the sodium salt of triciribine phosphate (TCN-P), a small-molecule inhibitor of Akt signaling used in cell signaling, oncology, and metabolic research. It is supplied as a high-purity solid suitable for in vitro and preclinical studies.
- High purity: 99.8%.
- Molecular formula: C13H16N6NaO7P.
- Molecular weight: 422.27 g·mol^-1.
- Appearance: white to light yellow solid.
- Pack size: 25 MG.
- Storage: store under nitrogen, away from moisture; in solvent, -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.