Learn More
Cayman Chemical TrIclabendazole-13C-d3 500ug
SDP
Supplier:  Cayman Chemical 34011500 UG
Encompass_Preferred
An internal standard for the quantification of triclabendazole by GC- or LC-MS
Catalog No. 50-294-8103