Learn More
Medchemexpress LLC Triclabendazole sulfoxide | 100648-13-3 | 98.8% | 375.66 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY136450100MG
Encompass_Preferred
Triclabendazole sulfoxide (TCBZ-SO) is the main plasma metabolite of Triclabendazole. It exhibits anti-parasite effects and can inhibit the membrane transporter ABCG2/BCRP.
  • Main plasma metabolite of Triclabendazole.
  • Exhibits anti-parasite effects.
  • Inhibits membrane transporter ABCG2/BCRP.
  • Targets parasite, BCRP, and PDGFR.
  • Involved in anti-infection, membrane transporter/ion channel, and protein tyrosine kinase/RTK pathways.
Catalog No. 50-003-50280