Learn More
Cayman Chemical Tricosanoic Acid ethyl ester
SDP
Supplier:  Cayman Chemical 100068745 MG
Encompass_Preferred
A saturated fatty acid ethyl ester; can be used as a standard for analysis of lipid mixtures
Catalog No. 50-288-8755