Learn More
Medchemexpress LLC Tricyclo[3.3.1.13,7]decane-1-carboxamide, n-[2-[4-(diphenylmethyl)-1-piperazinyl]-2-oxoethyl]- | 554438-52-7 | 99.9% | 471.63 g/mol | C30H37N3O2 | 10MM 1ML

Supplier: Medchemexpress LLC HY12487110MM 1ML
LASV inhibitor 3.3 is a small-molecule inhibitor that interacts with Lassa fever virus glycoprotein and the host factor LAMP1 to affect viral membrane fusion and entry. It is supplied both as solid material and as a ready-to-use 10 mM solution in DMSO for research applications, and is characterized by a high-purity analytical certificate of analysis.
- Targets viral glycoprotein-mediated entry and host factor LAMP1.
- Available as solids and a 10 mM DMSO solution for convenient assay preparation.
- High purity (~99.9%) as shown by certificate of analysis.
- Recommended storage: powder at -20°C (long term) and solutions at -80°C (short term).
- Molecular formula C30H37N3O2 and molecular weight ~471.6 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.