Learn More
Medchemexpress LLC Tricyclohexylphosphine | 2622-14-2 | ≥99.0% | 280.43 | 25 G
Supplier: Medchemexpress LLC 25GHY34614
Tricyclohexylphosphine is a biological molecule intended for research use only and not for medical applications. It is supplied as a solid, white to off-white powder. It is recommended to store this product at -20°C under nitrogen for optimal stability.
- Biological molecule
- Solid, white to off-white appearance
- Store at -20°C under nitrogen