Learn More
Strem, An Ascensus Company CAS# 7650-88-6. 10g. Tricyclopentylphosphine, min. 95% (10wt% in hexanes). MFCD00269834
SDP
Supplier:  Strem, An Ascensus Company 15618110G
Encompass
CAS# 7650-88-6. 10g. Tricyclopentylphosphine, min. 95% (10wt% in hexanes). MFCD00269834. Molecular Weight: 238.35. Molecular Formula: (C5H9)3P. Color/form: colorless liq. Strem# 15-6181. http://www.strem.com/catalog/v/15-6181/
Catalog No. NC1857068