Learn More
Strem, An Ascensus Company CAS# 7650-88-6. 5g. Tricyclopentylphosphine, min. 95%. MFCD00269834
SDP
Supplier:  Strem, An Ascensus Company 1561805G
Encompass
CAS# 7650-88-6. 5g. Tricyclopentylphosphine, min. 95%. MFCD00269834. Molecular Weight: 238.35. Molecular Formula: (C5H9)3P. Color/form: colorless liq. Strem# 15-6180. http://www.strem.com/catalog/v/15-6180/
Catalog No. 50-901-10395