Learn More
Medchemexpress LLC Tridecanedioic acid | 505-52-2 | 99.9% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY128421R
Encompass_Preferred
Tridecanedioic acid, also known as Brassylic Acid, is a chemical substance with the formula C13H24O4 and a molecular weight of 244.33. It appears as a white to off-white solid. This product is stable under recommended storage conditions and is not classified as a hazardous substance or mixture.
  • High purity, with HPLC analysis showing 99.9%
  • Stable under recommended storage conditions
  • Supplied as a solid
  • Chemical formula C13H24O4
Catalog No. 50-003-42814