Learn More
Medchemexpress LLC Tridecanoic acid-d25 | 202529-03-1 | 10mg
SDP
Supplier:  Medchemexpress LLC HYY1718S110MG
Encompass_Preferred
Tridecanoic acid-d25 is the deuterium labeled Tridecanoic acid Tridecanoic acid (N-Tridecanoic acid) a 13-carbon medium-chain saturated fatty acid can serve as an antipersister and antibiofilm agent that may be applied to research bacterial infections Tridecanoic acid inhibits Escherichia coli persistence and biofilm formation[1]
Catalog No. 50-002-66327