Learn More
Medchemexpress LLC Triethanolamine borate | 283-56-7 | 99.92% | 500 G
SDP
Supplier:  Medchemexpress LLC HYD0902500G
Encompass_Preferred
Triethanolamine borate is a compound belonging to the class of borates. It is commonly used in the manufacture of personal care and cosmetics as an emulsifier, pH adjuster and buffer. Triethanolamine borate helps stabilize and thicken products such as lotions and creams and can improve their overall texture. It is also used in industrial applications such as the production of lubricants and metalworking fluids.
  • Functions as an emulsifier, pH adjuster, and buffer.
  • Stabilizes and thickens personal care and cosmetic products.
  • Improves the overall texture of lotions and creams.
  • Applied in industrial settings for lubricants and metalworking fluids.
Catalog No. 50-004-10200