Learn More
Medchemexpress LLC Triethyl citrate (standard) | 77-93-0 | 99.57% | 276.29 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW011602R100MG
Encompass_Preferred
Triethyl citrate (Standard) is an analytical standard that is an ester of citric acid. It is intended for research and analytical applications and can be used as a plasticizer for cellulosic plastic-based nanocomposites. As an analytical standard, it is a reference standard supplied for assay and is commonly employed in qualitative, quantitative, and methodological research experiments using techniques like HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Can be used as a plasticizer for cellulosic plastic-based nanocomposites.
  • Reference standard for assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-54955