Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Triethylamine 100 ml 100000110

Supplier:  Waters Corp 100000110

Encompass_Preferred

Triethylamine 100mL for use with PicoTag Amino Acid Analysis

Catalog No. 50-780-000


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.