Learn More
Medchemexpress LLC Triethylammonium bicarbonate | 15715-58-9 | MFCD00466938 | 163.21 g·mol⁻¹ | C7H17NO3 | 10 ML
SDP
Supplier:  Medchemexpress LLC HYW69462910ML
Encompass_Preferred
Triethylammonium bicarbonate buffer (1.0 M, pH 8.5) is a volatile aqueous buffer supplied as a ready-to-use solution for analytical and preparative workflows such as LC-MS and peptide/protein sample preparation. The 10 mL vial format is suitable for low-volume workflows and easy handling.
  • Concentration: 1.0 M, pH 8.5
  • Volatile buffer suitable for LC-MS and mass spectrometry sample cleanup
  • Ready-to-use liquid format for convenient handling
  • Small-volume packaging for low-volume workflows
  • Stable when stored under recommended conditions
Catalog No. 50-002-84334