Learn More
Medchemexpress LLC Triethylammonium bicarbonate | 15715-58-9 | MFCD00466938 | 98.0% | 163.21 g/mol | C7H17NO3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW6946291ML
Encompass_Preferred
Triethylammonium bicarbonate is supplied as a 1.0 M aqueous buffer adjusted to pH 8.5 and is used to maintain a stable near-alkaline environment for enzymatic reactions, proteomic sample preparation, and mass spectrometry workflows.
  • Maintains stable pH at 8.5 for enzymatic reactions and digestions.
  • Volatile buffer compatible with mass spectrometry sample preparation.
  • High concentration (1.0 M) provides strong buffering capacity.
  • Supplied as an aqueous solution, colorless to light yellow.
  • Available in small-volume aliquots suitable for low-volume workflows.
  • High listed purity (98.0%) suitable for analytical applications.
Catalog No. 50-002-84951