Learn More
Medchemexpress LLC Triethylammonium bicarbonate | 15715-58-9 | MFCD00466938 | 98.0% | 163.21 g/mol | C7H17NO3 | 5 ML
SDP
Supplier:  Medchemexpress LLC HYW6946295ML
Encompass_Preferred
Aqueous triethylammonium bicarbonate buffer (1.0 M, pH 8.5) supplied for laboratory biochemical use. It provides a stable near-alkaline buffering environment for enzymatic reactions and general buffer preparation.
  • 1.0 M concentration at pH 8.5.
  • Suitable for buffer preparation in enzymatic and biochemical assays.
  • Supplied as an aqueous solution in small-volume formats for laboratory use.
  • Molecular weight 163.21 g/mol; CAS 15715-58-9.
Catalog No. 50-002-84332