Learn More
Selleck Chemical LLC Triethylenediamine E2967-100MG
SDP
Supplier:  Selleck Chemical LLC E2967100MG
Encompass_Preferred
Triethylenediamine (1 4-Diazabicyclo[2 2 2]octane DABCO) is a caged tertiary diamine commonly used as a strong hindered amine base in chemical synthesis It is used in the synthesis of isoxazole derivatives via dehydration of primary nitro compounds in the presence of dipolarophiles It can also be employed as a complexing ligand and as a catalyst
Catalog No. 50-000-40714