Learn More
Selleck Chemical LLC Triethylenetetramine 2HCl
SDP
Supplier:  Selleck Chemical LLC S658525MG
Encompass_Preferred
Triethylenetetramine 2HCl a Cu(II)-selective chelator is commonly used for the treatment of Wilson s disease
Catalog No. 50-000-02054