Learn More
Medchemexpress LLC Triethylmethylammoni 100g
SDP
Supplier:  Medchemexpress LLC HYW101255100G
Encompass_Preferred
Triethylmethylammonium chloride is a quaternary ammonium salt belonging to the class of alkylammonium salts This compound is widely used as a phase transfer catalyst in organic synthesis to facilitate the transfer of reactants between immiscible phases In addition it can be used as an electrolyte in electrochemical devices a surfactant in emulsion polymerization and a corrosion inhibitor Its unique chemical properties make it an important ingredient in a variety of industrial applications including pharmaceuticals agrochemicals and materials science
Catalog No. 50-002-06908