Learn More
Strem, An Ascensus Company CAS# 617-86-7. 100g. Triethylsilane, 99%. MFCD00009018
SDP
Supplier:  Strem, An Ascensus Company 147750100G
Encompass
CAS# 617-86-7. 100g. Triethylsilane, 99%. MFCD00009018. Molecular Weight: 116.28. Molecular Formula: (C2H5)3SiH. Color/form: colorless liq. Strem# 14-7750. http://www.strem.com/catalog/v/14-7750/
Catalog No. 50-901-09620