Learn More
Medchemexpress LLC Triflumizole | 68694-11-1 | >=90.0% - <=100.0% | 345.75 g/mol | C15H15ClF3N3O | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW020777
Encompass_Preferred
Triflumizole is an imidazole-class fungicide that inhibits ergosterol biosynthesis and is used to control powdery mildew and scab on a variety of fruits and agricultural crops. The substance has the chemical formula C15H15ClF3N3O and a molecular weight of 345.75 g/mol. It is supplied both as technical material and in formulated crop-protection products; concentration varies by formulation. Follow the product safety data sheet for handling, storage, and regulatory guidance.
  • Inhibits ergosterol biosynthesis in fungal pathogens.
  • Effective control of powdery mildew and scab on fruits and crops.
  • Available as technical grade and in formulated products with varying active concentrations.
  • Chemical formula C15H15ClF3N3O; molecular weight 345.75 g/mol.
  • Refer to the safety data sheet for handling, storage, and personal protective equipment.
Catalog No. 50-003-03343