Learn More
Medchemexpress LLC Triflumuron | 64628-44-0 | 99.0% | 100 G
SDP
Supplier:  Medchemexpress LLC HYB2049100G
Encompass_Preferred
Triflumuron is a potent chitin synthesis inhibitor that shows efficacy in emergence inhibition (EI) against *Culex quinquefasciatus* and *Aedes albopictus*, with EI50 values of 5.28 and 1.59 μg/L, respectively. It can be used as an insecticide and represents a potential tool for controlling disease vectors in public health.
  • Potent chitin synthesis inhibitor.
  • Shows efficacy in emergence inhibition against Culex quinquefasciatus and Aedes albopictus.
  • Can be used as an insecticide.
  • Potential tool for controlling disease vectors in public health.
Catalog No. 50-004-10997