Learn More
Selleck Chemical LLC Trifluoperazine
SDP
Supplier:  Selleck Chemical LLC S585625MG
Encompass_Preferred
Trifluoperazine (NSC-17474 RP-7623 SKF-5019) is an FDA-approved antipsychotic drug for schizophrenia It is an inhibitor of calmodulin (CaM) and Dopamine D2 receptor with an IC50 value of 1 2 nM for D2 receptor
Catalog No. 50-000-03903