Learn More
Apexbio Technology LLC Trifluoperazine 2HCl, 500mg Cas# 440-17-5
SDP
Supplier:  Apexbio Technology LLC B1397500
Encompass_Preferred
Trifluoperazine is a dopamine D2 receptor inhibitor with IC50 of 1.1 nM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8539