Learn More
Selleck Chemical LLC Trifluoperazine 5mg
SDP
Supplier:  Selleck Chemical LLC S58565MG
Encompass_Preferred
Trifluoperazine (NSC-17474, RP-7623, SKF-5019) is an FDA-approved antipsychotic drug for schizophrenia. It is an inhibitor of calmodulin (CaM) and Dopamine D2 receptor with an IC50 value of 1.2 nM for D2 receptor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3168