Learn More
Medchemexpress LLC Trifluoperazine dihydrochloride | 440-17-5 | MFCD00012656 | 99.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0532A500MG
Encompass_Preferred
Trifluoperazine dihydrochloride is an antipsychotic agent that functions by blocking central dopamine receptors. It is also a potent α1-adrenergic receptor antagonist and a potent NUPR1 inhibitor, exhibiting anticancer activity. Additionally, it acts as a calmodulin inhibitor and inhibits P-glycoprotein. This compound can be used in schizophrenia research and is a reversible inhibitor of influenza virus morphogenesis.
  • Antipsychotic agent
  • Blocks central dopamine receptors
  • Potent α1-adrenergic receptor antagonist
  • Potent NUPR1 inhibitor
  • Exerts anticancer activity
  • Calmodulin inhibitor
  • Inhibits P-glycoprotein
  • Used for schizophrenia research
  • Reversible inhibitor of influenza virus morphogenesis
Catalog No. 50-004-05850