Learn More
Medchemexpress LLC Trifluoperazine N-glucuronide-d3 (UGT1A4-d3) | 165602-90-4 | MFCD16294873 | 93.0% | 586.64 | 1 MG
Supplier: Medchemexpress LLC HY137083S1MG
Trifluoperazine N-glucuronide-d3 is deuterium labeled Trifluoperazine N-Glucuronide. Trifluoperazine N-Glucuronide (UGT1A4), a human UGT1A isoform expressed in the liver, catalyzes the imipramine and trifluoperazine N-glucuronide formation. It is for research use only and not sold to patients.
- Used as a tracer.
- Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.