Learn More
Strem, An Ascensus Company CAS# 407-25-0. 250g. Trifluoracetic anhydride, min. 98%. MFCD00000416
SDP
Supplier:  Strem, An Ascensus Company 097170250G
Encompass
CAS# 407-25-0. 250g. Trifluoracetic anhydride, min. 98%. MFCD00000416. Molecular Weight: 210.03. Molecular Formula: (CF3CO)2O. Color/form: colorless liq. Strem# 09-7170. http://www.strem.com/catalog/v/09-7170/
Catalog No. 50-901-09426