Learn More
TARGETMOL CHEMICALS INC TRIFLUOROMETHYL-TUBERCID 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T677535MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Trifluoromethyl-tubercidin (TFMT) inhibits host MTr1 and suppresses virus replication. TFMT inhibits MTr1 through interaction at its S-adenosyl-l-methionine binding pocket to restrict influenza virus replication. TFMT was effective in inhibiting viral replication in mice displayed little toxicity. purity: 99%
Catalog No. 50-257-9263