Learn More
Medchemexpress LLC Trifluperidol hydrochloride | 2062-77-3 | MFCD00055195 | 99.4% | 445.88 g/mol | C22H24ClF4NO2 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1244625MG
Encompass_Preferred
Trifluperidol hydrochloride is the hydrochloride salt of trifluperidol, a butyrophenone-class research compound used to study psychosis and sigma receptor pharmacology; it acts as a sigma-1 receptor antagonist (Ki = 3.3 nM), is supplied as a white to off-white solid, and has a typical purity of 99.4%.
  • High purity (99.4%) suitable for research applications.
  • Sigma-1 receptor antagonist with Ki = 3.3 nM.
  • White to off-white solid, suitable for laboratory handling.
  • Soluble in DMSO (40 mg/mL); ultrasonic and warming recommended.
  • Store sealed and away from moisture; in solution: -80°C up to 6 months, -20°C up to 1 month.
  • Available in small research pack sizes for assay and dose-finding studies.
Catalog No. 50-002-38906