Learn More
Medchemexpress LLC HY-N0616 5mg Medchemexpress, Trifolirhizin CAS:6807-83-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN06165MG
Encompass_Preferred
Medchemexpress, HY-N0616 5mg Trifolirhizin CAS:6807-83-6 Trifolirhizin is a pterocarpan flavonoid isolated from the roots of Sophora flavescens. Trifolirhizin possesses potent tyrosinase inhibitory activity with an IC50 of 506 μM. Trifolirhizin exhibits potential anti-inflammatory and anticancer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0296