Learn More
Medchemexpress LLC TriGalNAc CBz | 186613-57-0 | 98.7% | 633.64 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY147354
Encompass_Preferred
TriGalNAc CBz is a GalNAc derivative and a tri-GalNAc asialoglycoprotein receptor (ASGPR) ligand. It can be used for mRNA drug delivery as well as lysosomal targeted chimerism (LYTAC) studies. It acts as a linker for the synthesis of GalNAc-LYTACs, binding to the ASGPR receptor in hepatocytes to transport the target protein and receptor complex to the endosome, where tri-GalNAc is released in the acidic environment for target protein degradation.
  • GalNAc derivative
  • ASGPR ligand
  • Used for mRNA drug delivery
  • Used for lysosomal targeted chimerism (LYTAC) studies
  • Acts as a linker for synthesis of GalNAc-LYTACs
  • Binds specifically to the ASGPR receptor expressed in hepatocytes
  • Transports target protein and receptor complex to the endosome
Catalog No. 50-003-62910