Learn More
Medchemexpress LLC TriGalNAc CBz | 186613-57-0 | 98.73% | 633.64 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY147354100MG
Encompass_Preferred
TriGalNAc CBz is a GalNAc derivative and a tri-GalNAc asialoglycoprotein receptor (ASGPR) ligand. It is used in mRNA drug delivery and lysosomal targeted chimerism (LYTAC) studies.
  • Acts as a linker for the synthesis of GalNAc-LYTACs
  • Binds specifically to the ASGPR receptor in hepatocytes
  • Transports target protein and receptor complex to the endosome
  • Facilitates degradation of target protein in the lysosome
Catalog No. 50-003-59635