Learn More
Medchemexpress LLC TrigalNAc CBz | 186613-57-0 | 98.73% | 633.64 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14735450MG
Encompass_Preferred
TriGalNAc CBz is a GalNAc derivative and an asialoglycoprotein receptor (ASGPR) ligand. It can be utilized for mRNA drug delivery and lysosomal targeted chimerism (LYTAC) studies. It functions as a linker for the synthesis of GalNAc-LYTACs, specifically binding to the ASGPR receptor in hepatocytes to transport the target protein and receptor complex to the endosome for degradation.
  • GalNAc derivative
  • ASGPR ligand
  • Can be used for mRNA drug delivery
  • Can be used for lysosomal targeted chimerism (LYTAC) studies
  • Acts as a linker for the synthesis of GalNAc-LYTACs
  • Binds specifically to the ASGPR receptor in hepatocytes
  • Transports target protein and receptor complex to the endosome
  • Enables target protein transport to the lysosome for degradation
Catalog No. 50-003-60374