Learn More
Essendant TRIGGERCHMRSST9 7/8YL
Supplier:  Essendant IMP60192491
Encompass_Preferred
The Vitron rubber o-ring withstands the harshest chemicals preventing leakage of important and costly chemicals Chemical-resistant polymers that make up the trigger sprayer prevent degradation so the trigger sprayer continues to perform when used with the most severe chemicals The 28/400 mm thread finish is designed to fit the most popular bottles and containers Sprayer/Hose/Nozzle Type- Sprayer For Use With- 32 oz Bottles Material(s)- Polypropylene Color(s)- Yellow/White
Catalog No. 50-000-33712