Learn More
Cayman Chemical Triglyceride Colorimetric Assay Kit, 96 wells
SDP
Supplier:  Cayman Chemical 1001030396WELLS
Encompass_Preferred
The measurement of TG levels, in conjunction with other lipid assays, are useful in the diagnosis of primary and secondary hyperlipoproteinemia, dyslipidemia, and triglyceridemia. Cayman’s TG Assay Kit provides a simple, reproducible, and sensitive tool for assaying TGs in plasma, serum, cell lysates, and tissue homogenate samples. The assay is initiated with the enzymatic hydrolysis of the TGs by lipase to produce glycerol and free fatty acids. The glycerol released is subsequently measured by a coupled enzymatic reaction system with a colorimetric readout at 540 nm.
Catalog No. NC9656167