Learn More
Medchemexpress LLC Triglycidyl isocyanurate | 2451-62-9 | 99.6% | 297.26 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01143410G
Encompass_Preferred
Triglycidyl isocyanurate (TGIC; Teroxirone) is a triazene triepoxide with antiangiogenic and antineoplastic activities. It inhibits the growth of non-small-cell-lung cancer cells via p53 activation and induces cell apoptosis. Triglycidyl isocyanurate can be used for cancer research.
  • Antiangiogenic and antineoplastic activities
  • Inhibits the growth of non-small-cell-lung cancer cells via p53 activation
  • Induces cell apoptosis
  • Can be used for cancer research
Catalog No. 50-004-55017