Learn More
Strem, An Ascensus Company CAS# 258864-54-9. 50g. Trihexyl(tetradecyl)phosphonium chloride, min. 95% CYPHOS IL 101. MFCD04112746
SDP
Supplier:  Strem, An Ascensus Company 15638250G
Encompass
CAS# 258864-54-9. 50g. Trihexyl(tetradecyl)phosphonium chloride, min. 95% CYPHOS IL 101. MFCD04112746. Molecular Weight: 519.31. Molecular Formula: [(C6H13)3(C14H29)P]+Clae. Color/form: colorless liq. Strem# 15-6382. http://www.strem.com/catalog/v/15-6382
Catalog No. 50-901-10429